Assay Details
Target Gene Details
Entrez Gene ID: | 389860 |
Gene Name: | PAGE family member 2B |
Gene Aliases: |
CT16.5, GAGEE3, PAGE-2B |
Location: |
Chr.X:55027927-55078903 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PAGE2B | XM_011530785.2 | XP_011529087.1 | ||
XM_011530786.2 | XP_011529088.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion | SNORA109 SNORA11 LOC100421746 PFKFB1 USP51 FAM156B FGD1 PAGE5 MAGED2 FOXR2 MIR6857 HUWE1 MIR4536-2 TRO RIBC1 MIR98 IQSEC2 PAGE2B XAGE3 APEX2 KDM5C MTRNR2L10 PAGE3 MIR4536-1 MAGEH1 ALAS2 FAM104B FAM120C GPR173 FAM156A HSD17B10 ITIH6 MIR6895 WNK3 GNL3L KANTR TSR2 PHF8 PAGE2 MIR6894 TSPYL2 MIRLET7F2 SMC1A |
nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion | SNORA109 SNORA11 LOC100421746 PFKFB1 USP51 FAM156B FGD1 PAGE5 MAGED2 FOXR2 MIR6857 HUWE1 MIR4536-2 TRO RIBC1 MIR98 IQSEC2 PAGE2B XAGE3 APEX2 KDM5C MTRNR2L10 PAGE3 MIR4536-1 MAGEH1 ALAS2 FAM104B FAM120C GPR173 FAM156A HSD17B10 ITIH6 MIR6895 WNK3 GNL3L KANTR TSR2 PHF8 PAGE2 MIR6894 TSPYL2 MIRLET7F2 SMC1A |