Assay Details
Target Gene Details
Entrez Gene ID: | 203522 |
Gene Name: | integrator complex subunit 6 like |
Gene Aliases: |
DDX26B |
Location: |
Chr.X:135520630-135582535 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
INTS6L | NM_182540.4 | NP_872346.3 | ||
XM_005262394.2 | XP_005262451.1 | |||
XM_006724740.2 | XP_006724803.1 | |||
XM_006724741.3 | XP_006724804.1 | |||
XM_011531311.2 | XP_011529613.1 | |||
XM_017029342.1 | XP_016884831.1 | |||
XM_017029343.1 | XP_016884832.1 | |||
XM_017029344.1 | XP_016884833.1 | |||
XM_017029345.1 | XP_016884834.1 | |||
XM_017029346.1 | XP_016884835.1 | |||
XM_017029347.1 | XP_016884836.1 | |||
XM_017029348.1 | XP_016884837.1 | |||
XM_017029349.1 | XP_016884838.1 | |||
AK096544.1 | BAC04813.1 | |||
AK308423.1 | ||||
AL833319.1 | ||||
BC140715.1 | ||||
BX648113.1 | ||||
CN343659.1 | ||||
CR936620.1 | CAI56763.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758895 | Chr.X:135468612 - 136072582 on Build GRCh38 | Gain+Loss | CT45A9 SLC9A6 CT45A1 CT45A2 INTS6L SAGE1 CT45A10 INTS6L-AS1 CT45A5 CT45A3 CT45A7 CT45A8 CT45A6 MMGT1 |
nsv522205 | Chr.X:135374609 - 135533826 on Build GRCh38 | Gain | INTS6L-AS1 INTS6L LOC100506790 SMIM10L2A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |