Assay Details
Target Gene Details
Entrez Gene ID: | 100874530 |
Gene Name: | APOBEC3B antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.22:38991559-38998220 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APOBEC3B-AS1 | NR_104187.1 | |||
CX754295.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3627 | Chr.22:38952080 - 39000579 on Build GRCh38 | Deletion | APOBEC3B APOBEC3B-AS1 APOBEC3A |
nsv1061176 | Chr.22:38967602 - 38999490 on Build GRCh38 | Gain+Loss | APOBEC3B APOBEC3B-AS1 |
esv2760711 | Chr.22:38955461 - 39003307 on Build GRCh38 | Loss | APOBEC3B APOBEC3B-AS1 APOBEC3A |
dgv172n111 | Chr.22:38947911 - 39007789 on Build GRCh38 | Deletion | APOBEC3B APOBEC3B-AS1 APOBEC3A |
esv3647751 | Chr.22:38997523 - 39050430 on Build GRCh38 | Gain | APOBEC3C APOBEC3F APOBEC3B-AS1 APOBEC3D |
nsv198 | Chr.22:38954302 - 39000579 on Build GRCh38 | Deletion | APOBEC3B APOBEC3B-AS1 APOBEC3A |
nsv834196 | Chr.22:38996564 - 39214999 on Build GRCh38 | Loss | APOBEC3C APOBEC3G CBX7 APOBEC3F APOBEC3B-AS1 APOBEC3D APOBEC3H |