Assay Details
Target Gene Details
Entrez Gene ID: | 1415 |
Gene Name: | crystallin beta B2 |
Gene Aliases: |
CCA2, CRYB2, CRYB2A, CTRCT3, D22S665 |
Location: |
Chr.22:25211660-25231869 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CRYBB2 | XM_006724141.3 | 1 | 102 | XP_006724204.1 |
XM_011529900.2 | 1 | 102 | XP_011528202.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv469659 | Chr.22:25202943 - 25366882 on Build GRCh38 | Gain+Loss | LOC100652901 CRYBB2 IGLL3P LOC105372969 LRP5L CRYBB3 |
nsv834162 | Chr.22:25149393 - 25313043 on Build GRCh38 | Loss | KIAA1671 CRYBB2 LOC105372969 CRYBB3 |
nsv9888 | Chr.22:25204810 - 25247101 on Build GRCh38 | Gain | CRYBB2 CRYBB3 |
dgv24e196 | Chr.22:25116284 - 25403903 on Build GRCh38 | Duplication | LOC100652901 KIAA1671 CRYBB2 IGLL3P LOC105372969 LRP5L CRYBB3 |
esv2758829 | Chr.22:24970834 - 25650546 on Build GRCh38 | Gain+Loss | LOC100652901 KIAA1671 CRYBB2P1 CRYBB2 IGLL3P MIR6817 LOC105372966 LOC105372969 LRP5L LOC100128531 CRYBB3 GRK3 |
nsv428388 | Chr.22:25153850 - 25435228 on Build GRCh38 | Loss | LOC100652901 KIAA1671 CRYBB2 IGLL3P LOC105372969 LRP5L CRYBB3 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |