Assay Details
Target Gene Details
Entrez Gene ID: | 57553 |
Gene Name: | microtubule associated monooxygenase, calponin and LIM domain containing 3 |
Gene Aliases: |
MICAL-3 |
Location: |
Chr.22:17787649-18024559 on Build GRCh38 |
Assay Gene Location: | Within Intron 38 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509806 | Chr.22:17806210 - 17883749 on Build GRCh38 | Insertion | MICAL3 |
nsv526826 | Chr.22:17411511 - 17836413 on Build GRCh38 | Gain | LOC101929372 MIR3198-1 BCL2L13 LINC00528 BID SLC25A18 CECR2 ATP6V1E1 MICAL3 |
nsv834120 | Chr.22:17733890 - 17904438 on Build GRCh38 | Loss | MIR3198-1 LINC00528 BID MICAL3 |
esv3893428 | Chr.22:17578563 - 18040448 on Build GRCh38 | Loss | LOC101929372 MIR3198-1 BCL2L13 FLJ41941 LINC00528 BID MIR648 SLC25A18 ATP6V1E1 MICAL3 |
nsv428385 | Chr.22:17491941 - 18886703 on Build GRCh38 | Gain+Loss | LOC729461 RIMBP3 TUBA8 MICAL3 LOC100996401 LOC102725072 LINC00528 BID MIR648 PI4KAP1 LOC101929372 PEX26 BCL2L13 LOC100996415 LOC100996432 FAM230A GGT3P MIR3198-1 FLJ41941 SLC25A18 CECR2 TMEM191B ATP6V1E1 USP18 LOC642643 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |