Assay Details
Target Gene Details
Entrez Gene ID: | 54584 |
Gene Name: | G protein subunit beta 1 like |
Gene Aliases: |
DGCRK3, FKSG1, GY2, WDR14, WDVCF |
Location: |
Chr.22:19788411-19854939 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GNB1L | NM_053004.2 | NP_443730.1 | ||
AB051432.1 | ||||
AF238328.1 | AAG36826.1 | |||
AF301895.1 | AAG53933.1 | |||
AY007378.1 | AAG12162.1 | |||
BC012060.1 | AAH12060.1 | |||
CR456496.1 | CAG30382.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834129 | Chr.22:19705617 - 19919068 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv517478 | Chr.22:19703979 - 19817740 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
esv3893434 | Chr.22:19470889 - 20343372 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv828939 | Chr.22:19696032 - 20324465 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv828938 | Chr.22:19691069 - 20324465 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv471181 | Chr.22:19729797 - 19815256 on Build GRCh38 | Loss |
![]() ![]() |
esv3568260 | Chr.22:19777375 - 19881883 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
esv3647279 | Chr.22:19719848 - 19878980 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv7983n54 | Chr.22:19762002 - 20321870 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3575418 | Chr.22:18161776 - 19963846 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv588216 | Chr.22:19086099 - 19839117 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)