Assay Details
Target Gene Details
Entrez Gene ID: | 25829 |
Gene Name: | transmembrane protein 184B |
Gene Aliases: |
C22orf5, FM08, HS5O6A, HSPC256 |
Location: |
Chr.22:38216045-38273034 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM184B | XM_011530116.2 | XP_011528418.2 | ||
XM_017028758.1 | XP_016884247.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834194 | Chr.22:38063495 - 38240504 on Build GRCh38 | Loss | BAIAP2L2 TMEM184B PICK1 MAFF SNORA92 SLC16A8 PLA2G6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |