Assay Details
Target Gene Details
Entrez Gene ID: | 80833 |
Gene Name: | apolipoprotein L3 |
Gene Aliases: |
APOLIII, CG121, CG12_1, apoL-III |
Location: |
Chr.22:36140323-36166177 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv8093n54 | Chr.22:36137894 - 36187036 on Build GRCh38 | Loss | APOL3 |
esv2751964 | Chr.22:36137737 - 36183513 on Build GRCh38 | Loss | APOL3 |
dgv4563n100 | Chr.22:36142329 - 36186502 on Build GRCh38 | Loss | APOL3 |
esv3647677 | Chr.22:36137690 - 36164506 on Build GRCh38 | Loss | APOL3 |
nsv526454 | Chr.22:36143804 - 36187036 on Build GRCh38 | Loss | APOL3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |