Assay Details
Target Gene Details
Entrez Gene ID: | 5152 |
Gene Name: | phosphodiesterase 9A |
Gene Aliases: |
HSPDE9A2 |
Location: |
Chr.21:42653752-42775509 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3534 | Chr.21:42650074 - 42681450 on Build GRCh38 | Insertion | PDE9A LOC101928255 |
nsv522001 | Chr.21:42421796 - 42827928 on Build GRCh38 | Gain | LOC101928212 PDE9A UBASH3A LOC101928255 LOC101928311 SLC37A1 LOC101928233 LOC101928284 RSPH1 LOC101930094 |
dgv2500e59 | Chr.21:42669673 - 42671171 on Build GRCh38 | Duplication | PDE9A |
esv2666871 | Chr.21:42615442 - 43467915 on Build GRCh38 | Deletion | NDUFV3 CRYAA SIK1 LINC00313 U2AF1 WDR4 PDE9A ERVH48-1 PKNOX1 CBS FRGCA LOC101928311 LOC400870 LINC00319 MIR5692B HSF2BP LINC00322 LOC101928255 TCONS_00029157 LOC101928284 |
nsv587655 | Chr.21:42665548 - 42680969 on Build GRCh38 | Loss | PDE9A |
esv3647086 | Chr.21:42641558 - 42685014 on Build GRCh38 | Gain | PDE9A LOC101928255 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |