Assay Details
Target Gene Details
Entrez Gene ID: | 754 |
Gene Name: | pituitary tumor-transforming 1 interacting protein |
Gene Aliases: |
C21orf1, C21orf3, PBF |
Location: |
Chr.21:44849585-44873903 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTTG1IP | NM_001286822.1 | NP_001273751.1 | ||
NM_004339.3 | NP_004330.1 | |||
NR_104597.1 | ||||
AF149785.1 | AAF73770.1 | |||
AK094882.1 | ||||
AK290139.1 | ||||
AK293318.1 | ||||
AK298557.1 | ||||
AK299204.1 | ||||
AK315626.1 | ||||
BC000415.2 | AAH00415.1 | |||
BC012858.1 | AAH12858.1 | |||
BC019295.1 | AAH19295.1 | |||
BC020983.1 | AAH20983.1 | |||
BC031097.1 | AAH31097.1 | |||
BC034250.1 | AAH34250.1 | |||
BG721942.1 | ||||
CN295711.1 | ||||
Z50022.1 | CAA90325.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1058550 | Chr.21:44807248 - 44898019 on Build GRCh38 | Loss | ITGB2 PTTG1IP SUMO3 |
esv3647136 | Chr.21:44802580 - 44905193 on Build GRCh38 | Loss | ITGB2 PTTG1IP SUMO3 LINC01424 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |