Assay Details
Target Gene Details
Entrez Gene ID: | 8208 |
Gene Name: | chromatin assembly factor 1 subunit B |
Gene Aliases: |
CAF-1, CAF-IP60, CAF1, CAF1A, CAF1P60, MPHOSPH7, MPP7 |
Location: |
Chr.21:36380145-36416827 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHAF1B | NM_005441.2 | NP_005432.1 | ||
XM_011529755.2 | XP_011528057.1 | |||
XM_017028477.1 | XP_016883966.1 | |||
XM_017028478.1 | XP_016883967.1 | |||
AK313152.1 | ||||
AL711314.1 | ||||
BC021218.2 | AAH21218.1 | |||
U20980.1 | AAA76737.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33578 | Chr.21:36116014 - 36447329 on Build GRCh38 | Loss | CHAF1B CBR3-AS1 DOPEY2 LOC100133286 CBR3 MORC3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |