Assay Details
Target Gene Details
Entrez Gene ID: | 114044 |
Gene Name: | MCM3AP antisense RNA 1 |
Gene Aliases: |
C21orf85, MCM3AP-AS, MCM3APAS, MCM3APASB, NCRNA00031 |
Location: |
Chr.21:46229231-46251701 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MCM3AP-AS1 | NR_002776.4 | |||
NR_110565.1 | ||||
NR_110566.1 | ||||
NR_110567.1 | ||||
AF426263.1 | ||||
AK001370.1 | ||||
BC059792.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv519904 | Chr.21:46163592 - 46461396 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv509804 | Chr.21:45879599 - 46235140 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv470910 | Chr.21:45933861 - 46665076 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1060978 | Chr.21:46068159 - 46413726 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)