Assay Details
Target Gene Details
Entrez Gene ID: | 5152 |
Gene Name: | phosphodiesterase 9A |
Gene Aliases: |
HSPDE9A2 |
Location: |
Chr.21:42653752-42775509 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522001 | Chr.21:42421796 - 42827928 on Build GRCh38 | Gain | LOC101928311 LOC101928212 RSPH1 LOC101928233 SLC37A1 LOC101928284 LOC101928255 LOC101930094 UBASH3A PDE9A |
esv2666871 | Chr.21:42615442 - 43467915 on Build GRCh38 | Deletion | MIR5692B NDUFV3 CBS LOC101928284 LOC101928255 U2AF1 SIK1 ERVH48-1 LOC400870 PDE9A LOC101928311 HSF2BP PKNOX1 LINC00319 WDR4 FRGCA LINC00322 TCONS_00029157 LINC00313 CRYAA |
esv3647086 | Chr.21:42641558 - 42685014 on Build GRCh38 | Gain | LOC101928255 PDE9A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |