Assay Details
Target Gene Details
Entrez Gene ID: | 8568 |
Gene Name: | ribosomal RNA processing 1 |
Gene Aliases: |
D21S2056E, NNP-1, NOP52, RRP1A |
Location: |
Chr.21:43789537-43804102 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RRP1 | NM_003683.5 | NP_003674.1 | ||
XM_017028485.1 | XP_016883974.1 | |||
XM_017028486.1 | XP_016883975.1 | |||
AK223185.1 | BAD96905.1 | |||
AK223215.1 | BAD96935.1 | |||
AK303000.1 | ||||
AK313116.1 | ||||
BC000380.2 | AAH00380.1 | |||
BC014787.1 | AAH14787.2 | |||
DA275615.1 | ||||
U79775.1 | AAC51625.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509799 | Chr.21:43724009 - 43852846 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() ![]() |
nsv834108 | Chr.21:43758156 - 43949760 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv834107 | Chr.21:43735444 - 43874665 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv953642 | Chr.21:43731220 - 43827719 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() |
nsv470904 | Chr.21:43731164 - 43838166 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv520305 | Chr.21:43756077 - 43826445 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
esv33817 | Chr.21:43749531 - 44647625 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)