Assay Details
Target Gene Details
Entrez Gene ID: | 5152 |
Gene Name: | phosphodiesterase 9A |
Gene Aliases: |
HSPDE9A2 |
Location: |
Chr.21:42653752-42775509 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3534 | Chr.21:42650074 - 42681450 on Build GRCh38 | Insertion | LOC101928255 PDE9A |
nsv522001 | Chr.21:42421796 - 42827928 on Build GRCh38 | Gain | SLC37A1 LOC101930094 LOC101928233 LOC101928311 LOC101928255 UBASH3A LOC101928212 LOC101928284 PDE9A RSPH1 |
esv2666871 | Chr.21:42615442 - 43467915 on Build GRCh38 | Deletion | SIK1 LINC00313 HSF2BP LOC101928311 LOC101928255 LOC400870 PKNOX1 CRYAA CBS LINC00322 PDE9A TCONS_00029157 ERVH48-1 NDUFV3 FRGCA MIR5692B LINC00319 WDR4 U2AF1 LOC101928284 |
nsv587655 | Chr.21:42665548 - 42680969 on Build GRCh38 | Loss | PDE9A |
esv3647086 | Chr.21:42641558 - 42685014 on Build GRCh38 | Gain | LOC101928255 PDE9A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |