Assay Details
Target Gene Details
Entrez Gene ID: | 6919 |
Gene Name: | transcription elongation factor A2 |
Gene Aliases: |
TFIIS |
Location: |
Chr.20:64042350-64072347 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TCEA2 | XM_011529022.1 | XP_011527324.1 | ||
XM_011529027.2 | XP_011527329.1 | |||
XM_017028036.1 | XP_016883525.1 | |||
XM_017028037.1 | XP_016883526.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv586727 | Chr.20:64048243 - 64052630 on Build GRCh38 | Gain |
![]() ![]() |
nsv470564 | Chr.20:64027492 - 64169828 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv586703 | Chr.20:63964886 - 64095388 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv509785 | Chr.20:63999103 - 64137376 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv428381 | Chr.20:63583375 - 64334167 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv953310 | Chr.20:64017648 - 64160147 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1160691 | Chr.20:63943650 - 64055199 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv586729 | Chr.20:64049159 - 64052630 on Build GRCh38 | Loss |
![]() ![]() |
esv2758807 | Chr.20:64006869 - 64334167 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)