Assay Details
Target Gene Details
Entrez Gene ID: | 22919 |
Gene Name: | microtubule associated protein RP/EB family member 1 |
Gene Aliases: |
EB1 |
Location: |
Chr.20:32819780-32850405 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAPRE1 | NM_012325.2 | NP_036457.1 | ||
XM_011528696.2 | XP_011526998.1 | |||
AK297273.1 | ||||
AK312590.1 | ||||
BC106068.1 | AAI06069.1 | |||
BC109281.3 | AAI09282.1 | |||
BC128442.1 | ||||
BJ992159.1 | ||||
U24166.1 | AAC09471.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1056411 | Chr.20:32469234 - 33355029 on Build GRCh38 | Loss | BPIFA2 BPIFB1 BPIFB6 BPIFA4P MAPRE1 COMMD7 LOC101929698 LOC149950 BPIFA3 BPIFB2 C20orf203 BPIFA1 BPIFB3 BPIFB4 SUN5 EFCAB8 LOC105372714 DNMT3B NOL4L |
nsv833959 | Chr.20:32829604 - 32982934 on Build GRCh38 | Loss | MAPRE1 EFCAB8 |
nsv833957 | Chr.20:32755316 - 32907992 on Build GRCh38 | Gain+Loss | MAPRE1 EFCAB8 DNMT3B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |