Assay Details
Target Gene Details
Entrez Gene ID: | 101927117 |
Gene Name: | PLCG1 antisense RNA 1 |
Gene Aliases: |
TOP1-AS1 |
Location: |
Chr.20:41097994-41138000 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLCG1-AS1 | NR_109889.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1064246 | Chr.20:40261343 - 42254320 on Build GRCh38 | Loss | PLCG1-AS1 EMILIN3 LOC100128988 CHD6 LPIN3 TOP1 MAFB ZHX3 PTPRT PLCG1 MIR6871 |
nsv833983 | Chr.20:41051961 - 41213015 on Build GRCh38 | Loss | PLCG1-AS1 TOP1 ZHX3 PLCG1 MIR6871 |