Assay Details
Target Gene Details
Entrez Gene ID: | 1789 |
Gene Name: | DNA methyltransferase 3 beta |
Gene Aliases: |
ICF, ICF1, M.HsaIIIB |
Location: |
Chr.20:32762385-32809356 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DNMT3B | NM_001207055.1 | NP_001193984.1 | ||
NM_001207056.1 | NP_001193985.1 | |||
NM_006892.3 | NP_008823.1 | |||
NM_175848.1 | NP_787044.1 | |||
NM_175849.1 | NP_787045.1 | |||
AB208880.1 | BAD92117.1 | |||
AF156487.1 | AAD53062.1 | |||
AF156488.1 | AAD53063.1 | |||
AF331857.1 | AAL57040.1 | |||
AK295176.1 | ||||
AK299776.1 | ||||
AK299821.1 | ||||
AK299915.1 | ||||
AK300857.1 | ||||
DQ321787.1 | ABC48951.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1056411 | Chr.20:32469234 - 33355029 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv833956 | Chr.20:32623070 - 32814357 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv833957 | Chr.20:32755316 - 32907992 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)