Assay Details
Target Gene Details
Entrez Gene ID: | 10732 |
Gene Name: | transcription factor like 5 |
Gene Aliases: |
CHA, E2BP-1, Figlb, bHLHe82 |
Location: |
Chr.20:62841014-62861763 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TCFL5 | NM_006602.3 | NP_006593.2 | ||
XM_005260185.3 | XP_005260242.1 | |||
XM_011528497.2 | XP_011526799.1 | |||
XM_011528499.1 | XP_011526801.1 | |||
AB012124.1 | BAA36557.1 | |||
AF070992.1 | AAD53986.1 | |||
AJ271337.1 | CAC24700.1 | |||
BC046933.1 | AAH46933.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834027 | Chr.20:62707655 - 62874914 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv828794 | Chr.20:62832958 - 62880035 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
nsv9825 | Chr.20:62775470 - 63601330 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv428380 | Chr.20:62632018 - 62935383 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3646297 | Chr.20:62847710 - 62848185 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)