Assay Details
Target Gene Details
Entrez Gene ID: | 100128496 |
Gene Name: | chromosome 20 open reading frame 78 |
Gene Aliases: |
dJ1068E13.1 |
Location: |
Chr.20:18809351-18830203 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C20orf78 | NM_001242671.2 | NP_001229600.1 | ||
NM_001302818.1 | NP_001289747.1 | |||
AW118618.1 | ||||
BX112808.1 | ||||
BX334499.2 | ||||
CR738909.1 | ||||
HY016900.1 | ||||
HY050710.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833934 | Chr.20:18778246 - 18958077 on Build GRCh38 | Loss | C20orf78 SCP2D1 LINC00652 LOC100270804 |