Assay Details
Target Gene Details
Entrez Gene ID: | 55653 |
Gene Name: | breast carcinoma amplified sequence 4 |
Gene Aliases: |
CNOL |
Location: |
Chr.20:50794894-50877736 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BCAS4 | NM_001010974.2 | NP_001010974.1 | ||
NM_017843.4 | NP_060313.3 | |||
NM_198799.3 | NP_942094.2 | |||
XM_011528886.2 | XP_011527188.1 | |||
XM_011528887.2 | XP_011527189.1 | |||
XM_017027932.1 | XP_016883421.1 | |||
AF361220.1 | AAL99633.1 | |||
AJ511266.1 | CAD53579.1 | |||
AK000502.1 | BAA91209.1 | |||
BC038381.1 | AAH38381.1 | |||
BC047337.1 | AAH47337.1 | |||
BP212127.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834002 | Chr.20:50810235 - 50986764 on Build GRCh38 | Loss | MOCS3 ADNP TMSB4XP6 BCAS4 DPM1 ADNP-AS1 |
nsv834001 | Chr.20:50625617 - 50820418 on Build GRCh38 | Loss | FAM65C PARD6B LOC100506175 BCAS4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |