Assay Details
Target Gene Details
Entrez Gene ID: | 64405 |
Gene Name: | cadherin 22 |
Gene Aliases: |
C20orf25, dJ998H6.1 |
Location: |
Chr.20:46173733-46308498 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDH22 | NM_021248.2 | NP_067071.1 | ||
XM_011528994.2 | XP_011527296.1 | |||
XM_011528995.1 | XP_011527297.1 | |||
AF035300.1 | AAB88183.1 | |||
AK289584.1 | ||||
BC136526.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1057686 | Chr.20:46105657 - 46434770 on Build GRCh38 | Loss | SLC35C2 CD40 LOC105372633 ELMO2 CDH22 |
esv2758796 | Chr.20:46064276 - 46336238 on Build GRCh38 | Loss | NCOA5 CD40 CDH22 |
nsv833993 | Chr.20:46091252 - 46290015 on Build GRCh38 | Gain | CD40 CDH22 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |