Assay Details
Target Gene Details
Entrez Gene ID: | 27134 |
Gene Name: | tight junction protein 3 |
Gene Aliases: |
ZO-3, ZO3 |
Location: |
Chr.19:3708337-3750813 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TJP3 | NM_001267560.1 | NP_001254489.1 | ||
AK091118.1 | ||||
AK304462.1 | ||||
AL555606.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509718 | Chr.19:3676947 - 3844683 on Build GRCh38 | Insertion | TJP3 APBA3 ZFR2 RAX2 MRPL54 PIP5K1C MATK |
nsv470114 | Chr.19:3372516 - 3855979 on Build GRCh38 | Loss | SMIM24 RAX2 FZR1 C19orf71 MRPL54 MFSD12 MATK DOHH CACTIN TJP3 CACTIN-AS1 APBA3 TBXA2R ZFR2 NFIC PIP5K1C HMG20B GIPC3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |