Assay Details
Target Gene Details
Entrez Gene ID: | 125965 |
Gene Name: | cytochrome c oxidase subunit 6B2 |
Gene Aliases: |
COXVIB2, CT59 |
Location: |
Chr.19:55349702-55354814 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
COX6B2 | NM_144613.4 | NP_653214.2 | ||
AK057427.1 | ||||
BC026123.2 | ||||
BC100899.2 | AAI00900.1 | |||
BC100900.1 | AAI00901.1 | |||
BC100901.1 | AAI00902.1 | |||
BC100902.1 | AAI00903.1 | |||
BG717038.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953624 | Chr.19:55302033 - 55625034 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv470162 | Chr.19:55334018 - 55390889 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv516199 | Chr.19:55342945 - 55389731 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)