Assay Details
Target Gene Details
Entrez Gene ID: | 6929 |
Gene Name: | transcription factor 3 |
Gene Aliases: |
AGM8, E2A, E47, ITF1, TCF-3, VDIR, bHLHb21 |
Location: |
Chr.19:1609290-1652590 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv32942 | Chr.19:1320268 - 1999205 on Build GRCh38 | Gain+Loss | ATP8B3 RPS15 ADAT3 MUM1 MBD3 DAZAP1 CSNK1G2 KLF16 PLK5 ADAMTSL5 ABHD17A PCSK4 MEX3D REXO1 ONECUT3 TCF3 BTBD2 GAMT C19orf25 NDUFS7 APC2 MIR1909 UQCR11 LOC100288123 REEP6 CSNK1G2-AS1 SCAMP4 |
nsv953942 | Chr.19:1371602 - 2057401 on Build GRCh38 | Deletion | ATP8B3 RPS15 ADAT3 MUM1 MBD3 DAZAP1 CSNK1G2 KLF16 PLK5 ADAMTSL5 ABHD17A PCSK4 MEX3D REXO1 ONECUT3 TCF3 BTBD2 GAMT C19orf25 NDUFS7 APC2 MKNK2 MIR1909 UQCR11 LOC100288123 REEP6 CSNK1G2-AS1 SCAMP4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |