Assay Details
Target Gene Details
Entrez Gene ID: | 51343 |
Gene Name: | fizzy/cell division cycle 20 related 1 |
Gene Aliases: |
CDC20C, CDH1, FZR, FZR2, HCDH, HCDH1 |
Location: |
Chr.19:3506115-3536757 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FZR1 | NM_016263.3 | NP_057347.2 | ||
XM_005259573.4 | XP_005259630.1 | |||
AF083810.1 | AAD52030.1 | |||
AF102507.1 | AAD26623.1 | |||
BC013413.2 | AAH13413.1 | |||
DB462260.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953948 | Chr.19:3171703 - 3700002 on Build GRCh38 | Deletion | TBXA2R PIP5K1C FZR1 CACTIN HMG20B DOHH NCLN GIPC3 C19orf71 SMIM24 MFSD12 NFIC CACTIN-AS1 CELF5 S1PR4 |
nsv470114 | Chr.19:3372516 - 3855979 on Build GRCh38 | Loss | TJP3 TBXA2R RAX2 MRPL54 PIP5K1C APBA3 FZR1 CACTIN HMG20B ZFR2 DOHH GIPC3 MATK C19orf71 SMIM24 MFSD12 NFIC CACTIN-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |