Assay Details
Target Gene Details
Entrez Gene ID: | 5585 |
Gene Name: | protein kinase N1 |
Gene Aliases: |
DBK, PAK-1, PAK1, PKN, PKN-ALPHA, PRK1, PRKCL1 |
Location: |
Chr.19:14433354-14471867 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PKN1 | NM_002741.3 | NP_002732.3 | ||
XM_011528128.1 | XP_011526430.1 | |||
AK313886.1 | ||||
BC040061.1 | AAH40061.1 | |||
BC094766.1 | AAH94766.1 | |||
D26181.1 | BAA05169.1 | |||
S75546.1 | AAB33345.1 | |||
U33053.1 | AAC50209.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470127 | Chr.19:14434730 - 14479800 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv1160590 | Chr.19:14398750 - 14462842 on Build GRCh38 | Deletion |
![]() ![]() ![]() |
nsv833764 | Chr.19:14381787 - 14504061 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)