Assay Details
Target Gene Details
Entrez Gene ID: | 79948 |
Gene Name: | phospholipid phosphatase related 3 |
Gene Aliases: |
LPPR3, LPR3, PRG-2, PRG2 |
Location: |
Chr.19:812488-824244 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLPPR3 | NM_001270366.1 | 8 | 1757 | NP_001257295.1 |
NM_024888.2 | 7 | 1841 | NP_079164.1 | |
XM_011528317.2 | 7 | 2433 | XP_011526619.1 | |
AF357888.1 | 7 | 2104 | AAO85401.1 | |
AF541282.1 | 7 | 1841 | AAP57771.1 | |
AK021597.1 | 4 | 958 | BAB13851.1 | |
AL136596.1 | 7 | 1841 | CAB66531.1 | |
AY304517.1 | 7 | 1841 | AAP72154.1 | |
AY436785.1 | 8 | 1756 | AAR10818.1 | |
BC012339.1 | AAH12339.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv566n67 | Chr.19:805448 - 814402 on Build GRCh38 | Gain | MIR3187 PTBP1 PLPPR3 |
dgv134n111 | Chr.19:607984 - 967584 on Build GRCh38 | Deletion | FSTL3 HCN2 MIR3187 PTBP1 CFD PLPPR3 PRTN3 POLRMT PALM RNF126 KISS1R ELANE RNU6-9 PRSS57 LOC101928450 FGF22 ARID3A AZU1 R3HDM4 MISP MED16 MIR4745 |
nsv578172 | Chr.19:811414 - 841398 on Build GRCh38 | Loss | MIR3187 PTBP1 PLPPR3 PRTN3 AZU1 |
nsv578167 | Chr.19:797079 - 850574 on Build GRCh38 | Loss | MIR3187 PTBP1 PLPPR3 PRTN3 AZU1 MIR4745 |
nsv828354 | Chr.19:796933 - 815173 on Build GRCh38 | Gain | MIR3187 PTBP1 PLPPR3 MIR4745 |
nsv953938 | Chr.19:355801 - 1104501 on Build GRCh38 | Deletion | HCN2 MIR3187 CFD CNN2 WDR18 PRTN3 POLRMT C2CD4C ODF3L2 GPX4 KISS1R ABCA7 LOC101928450 SHC2 ARID3A AZU1 RNU6-2 TMEM259 MED16 MIR4745 GZMM FSTL3 PTBP1 BSG LOC105372235 MADCAM1 PLPPR3 TPGS1 PALM RNF126 ELANE POLR2E RNU6-9 PRSS57 FGF22 THEG CDC34 GRIN3B R3HDM4 MISP ARHGAP45 |
nsv470101 | Chr.19:401714 - 1008505 on Build GRCh38 | Loss | HCN2 MIR3187 CFD WDR18 PRTN3 POLRMT C2CD4C ODF3L2 KISS1R LOC101928450 SHC2 ARID3A AZU1 MED16 MIR4745 GZMM FSTL3 PTBP1 BSG LOC105372235 MADCAM1 PLPPR3 TPGS1 PALM RNF126 ELANE RNU6-9 PRSS57 FGF22 CDC34 GRIN3B R3HDM4 MISP |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |