Assay Details
Target Gene Details
Entrez Gene ID: | 79143 |
Gene Name: | membrane bound O-acyltransferase domain containing 7 |
Gene Aliases: |
BB1, LENG4, LPIAT, LRC4, MBOA7, OACT7, hMBOA-7 |
Location: |
Chr.19:54173412-54190050 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MBOAT7 | NM_001146056.2 | NP_001139528.1 | ||
NM_001146082.2 | NP_001139554.1 | |||
NM_001146083.2 | NP_001139555.1 | |||
NM_024298.4 | NP_077274.3 | |||
XM_011527299.2 | XP_011525601.1 | |||
XM_011527300.2 | XP_011525602.1 | |||
XM_017027296.1 | XP_016882785.1 | |||
AK055908.1 | BAB71043.1 | |||
AK122789.1 | ||||
AK123290.1 | 1 | 497 | ||
AK293199.1 | ||||
AK298689.1 | ||||
AK311493.1 | ||||
BC002512.2 | AAH02512.2 | |||
BC003164.1 | AAH03164.2 | |||
BC006309.2 | AAH06309.1 | |||
BC015857.1 | ||||
BU633996.1 | ||||
EU016381.1 | ||||
S82470.1 | AAB37433.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9751 | Chr.19:54158885 - 54193528 on Build GRCh38 | Gain | TMC4 TSEN34 LENG1 MBOAT7 |
nsv953613 | Chr.19:54097214 - 54198733 on Build GRCh38 | Deletion | CNOT3 OSCAR TMC4 TSEN34 TFPT LENG1 MBOAT7 NDUFA3 PRPF31 |
esv2718812 | Chr.19:53184636 - 54322450 on Build GRCh38 | Deletion | MIR517A ZNF845 MIR372 MIR515-1 MIR519D CACNG6 ZNF761 MIR522 NDUFA3 TPM3P9 MIR520G MIR373 MIR519B MIR935 ZNF665 MIR519C ZNF525 LENG1 MIR518C MIR521-1 MIR512-1 BIRC8 LILRA6 PRKCG MIR523 LOC105372457 TMC4 MIR512-2 LILRB2 MIR516B1 MIR516A2 MIR524 MIR1283-2 MIR520E CACNG7 MIR526A1 MIR518B ZNF813 LOC284379 MIR520F OSCAR TARM1 TSEN34 TFPT LILRB3 MIR516A1 VN1R4 VSTM1 DPRX MIR520H MIR515-2 MIR527 ZNF331 MIR525 MIR520C MIR520B CACNG8 NLRP12 ZNF765 MIR518A1 ZNF818P MIR516B2 MIR518F MIR1323 LILRA5 ZNF677 RPS9 MIR1283-1 MIR517C MIR4752 MIR526B MIR518D MIR498 MIR518E MIR371A MBOAT7 VN1R2 PRPF31 MIR526A2 MIR517B MIR520D MYADM CNOT3 MIR521-2 MIR519E MIR520A MIR518A2 MIR519A2 FAM90A27P MIR519A1 MIR371B LILRB5 |
nsv510774 | Chr.19:54158011 - 54269511 on Build GRCh38 | Deletion | RPS9 LILRA6 TMC4 TSEN34 LILRB3 LENG1 MBOAT7 LILRB5 |
esv2758771 | Chr.19:54163847 - 54954956 on Build GRCh38 | Gain+Loss | LENG8-AS1 LILRA2 KIR3DL1 CDC42EP5 KIR3DX1 LILRP2 LILRB4 LENG8 LOC102724300 TTYH1 LILRA1 KIR2DL4 MIR8061 LILRA4 LILRA5 RPS9 LILRA6 LAIR2 FCAR MIR4752 TMC4 KIR3DL3 LOC101928804 MBOAT7 KIR2DL1 LILRB2 NLRP7 LENG9 LILRB1 KIR3DL2 KIR2DL3 TSEN34 LILRB3 LAIR1 NCR1 KIR2DS4 LILRB5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |