Assay Details
Target Gene Details
Entrez Gene ID: | 4782 |
Gene Name: | nuclear factor I C |
Gene Aliases: |
CTF, CTF5, NF-I, NFI |
Location: |
Chr.19:3359563-3469217 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFIC | NM_001245005.1 | NP_001231934.1 | ||
NM_205843.2 | NP_995315.1 | |||
XM_006722759.2 | XP_006722822.2 | |||
XM_017026836.1 | XP_016882325.1 | |||
AK289885.1 | ||||
AK304816.1 | ||||
BP279194.1 | ||||
BQ894051.1 | ||||
X12492.1 | CAA31012.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509717 | Chr.19:3271020 - 3392050 on Build GRCh38 | Insertion | NFIC CELF5 |
nsv1058546 | Chr.19:3321667 - 3369170 on Build GRCh38 | Gain | NFIC |
nsv953948 | Chr.19:3171703 - 3700002 on Build GRCh38 | Deletion | DOHH C19orf71 MFSD12 NFIC GIPC3 CACTIN-AS1 NCLN PIP5K1C S1PR4 SMIM24 FZR1 HMG20B TBXA2R CACTIN CELF5 |
nsv833712 | Chr.19:3230782 - 3365290 on Build GRCh38 | Loss | NFIC CELF5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |