Assay Details
Target Gene Details
Entrez Gene ID: | 23094 |
Gene Name: | signal induced proliferation associated 1 like 3 |
Gene Aliases: |
CTRCT45, SPAL3, SPAR3 |
Location: |
Chr.19:37906823-38208372 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SIPA1L3 | NM_015073.2 | NP_055888.1 | ||
XM_017026518.1 | XP_016882007.1 | |||
BC036863.1 | AAH36863.1 | |||
BC150620.1 | ||||
BM193478.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833820 | Chr.19:37699820 - 37923409 on Build GRCh38 | Loss | LOC644554 LOC100631378 LOC105372394 ZNF573 ZNF607 WDR87 SIPA1L3 |
nsv1064401 | Chr.19:37897157 - 37937344 on Build GRCh38 | Loss | WDR87 SIPA1L3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |