Assay Details
Target Gene Details
Entrez Gene ID: | 6929 |
Gene Name: | transcription factor 3 |
Gene Aliases: |
AGM8, E2A, E47, ITF1, TCF-3, VDIR, bHLHb21 |
Location: |
Chr.19:1609290-1652590 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv32942 | Chr.19:1320268 - 1999205 on Build GRCh38 | Gain+Loss | ABHD17A REXO1 CSNK1G2-AS1 MIR1909 ADAT3 UQCR11 APC2 PCSK4 C19orf25 ATP8B3 PLK5 KLF16 MEX3D ONECUT3 NDUFS7 MBD3 TCF3 RPS15 ADAMTSL5 CSNK1G2 SCAMP4 LOC100288123 MUM1 GAMT REEP6 BTBD2 DAZAP1 |
nsv953942 | Chr.19:1371602 - 2057401 on Build GRCh38 | Deletion | ABHD17A REXO1 CSNK1G2-AS1 MIR1909 ADAT3 UQCR11 APC2 PCSK4 C19orf25 ATP8B3 PLK5 KLF16 MEX3D ONECUT3 NDUFS7 MBD3 TCF3 RPS15 ADAMTSL5 MKNK2 CSNK1G2 SCAMP4 LOC100288123 MUM1 GAMT REEP6 BTBD2 DAZAP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |