Assay Details
Target Gene Details
Entrez Gene ID: | 5064 |
Gene Name: | paralemmin |
Gene Aliases: |
- |
Location: |
Chr.19:708953-748330 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PALM | NM_001040134.1 | NP_001035224.1 | ||
NM_002579.2 | NP_002570.2 | |||
XM_005259565.4 | XP_005259622.1 | |||
XM_017026850.1 | XP_016882339.1 | |||
AK299809.1 | ||||
AK315168.1 | ||||
BC032449.1 | AAH32449.1 | |||
DA521203.1 | ||||
Y14770.1 | CAB37400.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv134n111 | Chr.19:607984 - 967584 on Build GRCh38 | Deletion | LOC101928450 MIR4745 RNF126 PALM FSTL3 ELANE HCN2 PTBP1 POLRMT FGF22 MED16 R3HDM4 RNU6-9 ARID3A PLPPR3 AZU1 MIR3187 PRSS57 CFD MISP PRTN3 KISS1R |
nsv577978 | Chr.19:294526 - 782854 on Build GRCh38 | Gain | LOC101929060 LOC101928450 RNF126 CDC34 SHC2 PALM BSG ODF3L2 FSTL3 HCN2 MIER2 MADCAM1 POLRMT FGF22 C2CD4C PRSS57 TPGS1 MISP THEG GZMM |
nsv953938 | Chr.19:355801 - 1104501 on Build GRCh38 | Deletion | MIR4745 SHC2 PALM BSG ODF3L2 ELANE ABCA7 HCN2 GRIN3B POLRMT MED16 R3HDM4 RNU6-9 C2CD4C PRSS57 POLR2E CFD TPGS1 WDR18 MISP PRTN3 RNU6-2 GZMM LOC101928450 RNF126 CDC34 FSTL3 MADCAM1 PTBP1 FGF22 ARID3A PLPPR3 TMEM259 AZU1 MIR3187 GPX4 LOC105372235 CNN2 ARHGAP45 THEG KISS1R |
nsv470101 | Chr.19:401714 - 1008505 on Build GRCh38 | Loss | MIR4745 SHC2 PALM BSG ODF3L2 ELANE HCN2 GRIN3B POLRMT MED16 R3HDM4 RNU6-9 C2CD4C PRSS57 CFD TPGS1 WDR18 MISP PRTN3 GZMM LOC101928450 RNF126 CDC34 FSTL3 MADCAM1 PTBP1 FGF22 ARID3A PLPPR3 AZU1 MIR3187 LOC105372235 KISS1R |
esv3643362 | Chr.19:712133 - 716174 on Build GRCh38 | Loss | PALM |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |