Assay Details
Target Gene Details
Entrez Gene ID: | 208 |
Gene Name: | AKT serine/threonine kinase 2 |
Gene Aliases: |
HIHGHH, PKBB, PKBBETA, PRKBB, RAC-BETA |
Location: |
Chr.19:40230317-40285531 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AKT2 | NM_001243027.2 | NP_001229956.1 | ||
NM_001243028.2 | NP_001229957.1 | |||
NM_001626.5 | NP_001617.1 | |||
XM_011526614.1 | XP_011524916.1 | |||
XM_011526615.1 | XP_011524917.1 | |||
XM_011526616.1 | XP_011524918.1 | |||
XM_011526618.1 | XP_011524920.1 | |||
XM_011526619.1 | XP_011524921.1 | |||
XM_011526620.1 | XP_011524922.1 | |||
XM_011526622.2 | XP_011524924.1 | |||
XM_017026470.1 | XP_016881959.1 | |||
XM_017026471.1 | XP_016881960.1 | |||
AK294453.1 | ||||
AK304209.1 | ||||
AK308006.1 | ||||
AK308906.1 | ||||
AK314619.1 | ||||
BC022779.1 | AAH22779.1 | |||
BC032709.1 | ||||
BC063421.1 | AAH63421.1 | |||
BC120994.1 | ||||
BC120995.1 | ||||
BQ219902.1 | ||||
KU159425.1 | ||||
M77198.1 | AAA36585.1 | |||
M95936.1 | AAA58364.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1062755 | Chr.19:40231556 - 40252328 on Build GRCh38 | Loss | AKT2 |
esv3644345 | Chr.19:40203124 - 40288712 on Build GRCh38 | Gain | AKT2 MIR641 CNTD2 MAP3K10 TTC9B |
esv3644344 | Chr.19:40191059 - 40328585 on Build GRCh38 | Gain | AKT2 MIR641 CNTD2 MAP3K10 C19orf47 TTC9B |
esv2758761 | Chr.19:40106513 - 40333613 on Build GRCh38 | Gain | AKT2 MIR641 CNTD2 MAP3K10 C19orf47 TTC9B |
nsv1059094 | Chr.19:40239470 - 40252328 on Build GRCh38 | Gain | AKT2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |