Assay Details
Target Gene Details
Entrez Gene ID: | 11133 |
Gene Name: | kaptin, actin binding protein |
Gene Aliases: |
2E4, MRT41 |
Location: |
Chr.19:47475141-47486792 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KPTN | NM_001291296.1 | NP_001278225.1 | ||
NM_007059.3 | NP_008990.2 | |||
NR_111923.1 | ||||
XM_011526399.2 | XP_011524701.1 | |||
XM_017026226.1 | XP_016881715.1 | |||
XM_017026227.1 | XP_016881716.1 | |||
XM_017026228.1 | XP_016881717.1 | |||
XM_017026229.1 | XP_016881718.1 | |||
AF105369.1 | AAD39358.1 | |||
AK023996.1 | ||||
AK225419.1 | ||||
AK298672.1 | ||||
BC009249.2 | AAH09249.1 | |||
DR759729.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758765 | Chr.19:47354499 - 47522248 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1056887 | Chr.19:47481963 - 47527369 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv579888 | Chr.19:47428355 - 47658874 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)