Assay Details
Target Gene Details
Entrez Gene ID: | 113178 |
Gene Name: | secretory carrier membrane protein 4 |
Gene Aliases: |
SCAMP-4 |
Location: |
Chr.19:1905374-1926013 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SCAMP4 | NM_079834.2 | NP_524558.1 | ||
AK074586.1 | 5 | 898 | BAC11075.1 | |
AK074967.1 | BAC11322.1 | |||
AK092056.1 | BAC03797.1 | |||
AK307641.1 | ||||
AK307700.1 | ||||
AL832731.1 | ||||
BC011747.2 | AAH11747.1 | |||
BC016509.1 | AAH16509.1 | |||
BC062598.1 | AAH62598.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893156 | Chr.19:1766738 - 2115791 on Build GRCh38 | Loss | MKNK2 ONECUT3 REXO1 ATP8B3 AP3D1 KLF16 CSNK1G2 SCAMP4 LOC100288123 MIR1909 BTBD2 MOB3A ADAT3 IZUMO4 ABHD17A CSNK1G2-AS1 |
esv32942 | Chr.19:1320268 - 1999205 on Build GRCh38 | Gain+Loss | C19orf25 MEX3D REXO1 TCF3 REEP6 MUM1 APC2 CSNK1G2 LOC100288123 PCSK4 BTBD2 ADAT3 ONECUT3 RPS15 NDUFS7 ATP8B3 KLF16 PLK5 ADAMTSL5 GAMT UQCR11 DAZAP1 SCAMP4 MIR1909 ABHD17A CSNK1G2-AS1 MBD3 |
nsv470107 | Chr.19:1782799 - 2100347 on Build GRCh38 | Loss | MKNK2 REXO1 ATP8B3 KLF16 CSNK1G2 SCAMP4 LOC100288123 MIR1909 BTBD2 MOB3A ADAT3 IZUMO4 ABHD17A CSNK1G2-AS1 |
nsv953942 | Chr.19:1371602 - 2057401 on Build GRCh38 | Deletion | C19orf25 MEX3D REXO1 TCF3 REEP6 MUM1 APC2 CSNK1G2 LOC100288123 PCSK4 BTBD2 ADAT3 MKNK2 ONECUT3 RPS15 NDUFS7 ATP8B3 KLF16 PLK5 ADAMTSL5 GAMT UQCR11 DAZAP1 SCAMP4 MIR1909 ABHD17A CSNK1G2-AS1 MBD3 |
nsv1160561 | Chr.19:1884553 - 2003982 on Build GRCh38 | Deletion | CSNK1G2 SCAMP4 BTBD2 ADAT3 ABHD17A CSNK1G2-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |