Assay Details
Target Gene Details
Entrez Gene ID: | 147645 |
Gene Name: | V-set and immunoglobulin domain containing 10 like |
Gene Aliases: |
- |
Location: |
Chr.19:51331541-51342124 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
VSIG10L | NM_001163922.1 | NP_001157394.1 | ||
BC049212.1 | AAH49212.1 | |||
BC131493.1 |