Assay Details
Target Gene Details
Entrez Gene ID: | 160 |
Gene Name: | adaptor related protein complex 2 alpha 1 subunit |
Gene Aliases: |
ADTAA, AP2-ALPHA, CLAPA1 |
Location: |
Chr.19:49766923-49807114 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AP2A1 | NM_014203.2 | NP_055018.2 | ||
NM_130787.2 | NP_570603.2 | |||
XM_011526556.2 | XP_011524858.1 | |||
XM_011526557.2 | XP_011524859.1 | |||
AK124343.1 | ||||
AK302005.1 | ||||
AL136925.1 | CAB66859.1 | |||
BC014214.2 | AAH14214.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | RCN3 C19orf68 SNAR-B1 SNORD35B SNAR-A2 SNAR-A13 SNAR-A9 PLEKHA4 SNAR-A5 TMEM143 IL4I1 FUZ SNAR-G2 CA11 AKT1S1 SNORD32A PRR12 MIR4751 CGB7 SLC6A16 NUCB1-AS1 RUVBL2 VRK3 NUCB1 CCDC155 HSD17B14 SNRNP70 PNKP SULT2B1 ELSPBP1 MAMSTR NTF4 MIR4324 MIR4749 SNAR-A7 CABP5 PTOV1-AS1 PTH2 FLJ26850 LOC105447645 ATF5 RPL18 SEC1P FCGRT IRF3 PPFIA3 TBC1D17 SNAR-B2 MIR4750 GRWD1 FGF21 BAX CD37 CARD8 BSPH1 CPT1C PRMT1 NUP62 CGB8 IZUMO1 SNAR-D SNAR-A12 SNAR-C1 ADM5 MIR150 LOC100287477 PTOV1-AS2 GFY CCDC114 DBP C19orf73 RPL13A LHB KCNJ14 PLA2G4C SNAR-C4 LMTK3 FLT3LG SNAR-A4 MED25 SNAR-A6 SNORD34 CGB3 MIR6799 NOSIP AP2A1 LIG1 SIGLEC16 EMP3 FTL TRPM4 ZNF473 RRAS FAM83E SNAR-A10 KDELR1 GYS1 MIR6798 CGB2 PIH1D1 GRIN2D ZNF114 LOC101059948 MIR5088 CGB5 BCAT2 TULP2 SNAR-A14 SNAR-A8 LIN7B SNAR-C3 PPP1R15A PRRG2 PTOV1 SNAR-A3 BCL2L12 SLC17A7 FUT2 FUT1 SNAR-A1 SIGLEC11 RASIP1 DKKL1 SNORD35A LOC105372430 RPS11 DHDH SNORD33 SCAF1 SNAR-C5 MIR6800 SNAR-G1 SYNGR4 CYTH2 KCNA7 PLA2G4C-AS1 SPHK2 TEAD2 ALDH16A1 SPACA4 CGB1 NTN5 SNAR-A11 CARD8-AS1 SNAR-C2 TSKS LOC101928295 HRC |
nsv470150 | Chr.19:49618821 - 49856789 on Build GRCh38 | Gain | RRAS MIR6800 PTOV1 IRF3 ADM5 BCL2L12 MED25 FUZ MIR5088 PTOV1-AS2 MIR6799 AP2A1 MIR4749 PTOV1-AS1 CPT1C PRMT1 PRR12 SCAF1 TSKS |
dgv53n68 | Chr.19:49724393 - 49943224 on Build GRCh38 | Loss | MIR6800 PTOV1 TBC1D17 MED25 MIR4750 IL4I1 PNKP FUZ PTOV1-AS2 MIR6799 AP2A1 MIR4749 PTOV1-AS1 AKT1S1 NUP62 MIR4751 TSKS ATF5 |
nsv953600 | Chr.19:49555044 - 49938043 on Build GRCh38 | Deletion | PRRG2 PTOV1 IRF3 BCL2L12 TBC1D17 MED25 MIR4750 IL4I1 FUZ MIR6799 NOSIP AP2A1 CPT1C AKT1S1 PRMT1 PRR12 NUP62 MIR4751 SCAF1 RRAS MIR6800 ADM5 PNKP MIR5088 PTOV1-AS2 MIR4749 PTOV1-AS1 TSKS ATF5 |
nsv1160655 | Chr.19:49489110 - 49841830 on Build GRCh38 | Deletion | RCN3 RPL13A FCGRT PRRG2 IRF3 SNORD35B BCL2L12 MED25 FUZ SNORD34 SNORD35A MIR6799 NOSIP AP2A1 CPT1C SNORD32A RPS11 PRMT1 PRR12 SNORD33 SCAF1 RRAS MIR6800 ADM5 MIR150 MIR5088 PTOV1-AS1 TSKS |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |