Assay Details
Target Gene Details
Entrez Gene ID: | 5296 |
Gene Name: | phosphoinositide-3-kinase regulatory subunit 2 |
Gene Aliases: |
MPPH, MPPH1, P85B, p85, p85-BETA |
Location: |
Chr.19:18153178-18170533 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PIK3R2 | NM_005027.3 | NP_005018.1 | ||
NR_073517.1 | ||||
BC032647.1 | ||||
BC070082.1 | AAH70082.1 | |||
BC090249.1 | AAH90249.1 | |||
X80907.1 | CAA56868.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv2429 | Chr.19:18123369 - 18168571 on Build GRCh38 | Deletion | MAST3 PIK3R2 |
nsv1160592 | Chr.19:18100050 - 18194768 on Build GRCh38 | Deletion | MPV17L2 MAST3 PIK3R2 IFI30 |
nsv952420 | Chr.19:18117791 - 18476990 on Build GRCh38 | Deletion | GDF15 MPV17L2 LSM4 MIR3189 PGPEP1 RAB3A JUND PDE4C SSBP4 LRRC25 KIAA1683 LOC729966 MIR3188 MAST3 LOC102725254 PIK3R2 ISYNA1 ELL IFI30 |
nsv833774 | Chr.19:18102951 - 18184519 on Build GRCh38 | Loss | MAST3 PIK3R2 IFI30 |
nsv833773 | Chr.19:18042161 - 18192178 on Build GRCh38 | Loss | IL12RB1 MAST3 PIK3R2 IFI30 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |