Assay Details
Target Gene Details
Entrez Gene ID: | 83855 |
Gene Name: | Kruppel like factor 16 |
Gene Aliases: |
BTEB4, DRRF, NSLP2 |
Location: |
Chr.19:1852399-1863565 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLF16 | NM_031918.3 | NP_114124.1 | ||
AF327440.1 | AAK15698.1 | |||
CV809534.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3893156 | Chr.19:1766738 - 2115791 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2669985 | Chr.19:1852046 - 1864993 on Build GRCh38 | Deletion |
![]() |
esv32942 | Chr.19:1320268 - 1999205 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv470107 | Chr.19:1782799 - 2100347 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1160560 | Chr.19:1791100 - 1916393 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv578338 | Chr.19:1847699 - 1869095 on Build GRCh38 | Loss |
![]() ![]() |
nsv578337 | Chr.19:1808864 - 1904613 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv953942 | Chr.19:1371602 - 2057401 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2660894 | Chr.19:1854446 - 1872693 on Build GRCh38 | Deletion |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)