Assay Details
Target Gene Details
Entrez Gene ID: | 160 |
Gene Name: | adaptor related protein complex 2 alpha 1 subunit |
Gene Aliases: |
ADTAA, AP2-ALPHA, CLAPA1 |
Location: |
Chr.19:49766923-49807114 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AP2A1 | NM_014203.2 | NP_055018.2 | ||
NM_130787.2 | NP_570603.2 | |||
XM_011526556.2 | XP_011524858.1 | |||
XM_011526557.2 | XP_011524859.1 | |||
AK124343.1 | ||||
AK302005.1 | ||||
AL136925.1 | CAB66859.1 | |||
BC014214.2 | AAH14214.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | SNAR-C1 PIH1D1 FUT1 SNAR-A3 RPS11 SNAR-A1 MIR150 CA11 GRWD1 MED25 SNAR-A4 C19orf73 CGB2 SCAF1 PRR12 DBP ZNF114 HSD17B14 SNAR-A11 CGB1 EMP3 SNAR-A13 PTH2 TSKS KCNA7 ALDH16A1 SNORD33 SIGLEC16 SNAR-G1 PPFIA3 AKT1S1 SNAR-A10 PLEKHA4 SNAR-C5 TRPM4 AP2A1 SNAR-A12 MIR4324 CCDC155 SPACA4 PRRG2 LMTK3 SNORD32A CARD8-AS1 SLC6A16 PTOV1-AS2 FCGRT C19orf68 IZUMO1 RPL13A RPL18 PTOV1-AS1 SNAR-G2 NUP62 KCNJ14 MIR6799 SNAR-A7 MAMSTR CGB5 SNAR-A8 SULT2B1 FUZ BSPH1 KDELR1 SNAR-A14 SNAR-A5 RUVBL2 PNKP SEC1P LOC101059948 NTF4 LIG1 ZNF473 MIR6800 MIR4750 RCN3 SIGLEC11 NUCB1 SNAR-D FUT2 SNORD34 TEAD2 LOC105372430 CABP5 GYS1 LOC101928295 PRMT1 GFY PPP1R15A FLJ26850 SNORD35B PLA2G4C SNAR-A6 SNAR-B1 RRAS GRIN2D PTOV1 LOC105447645 TULP2 NUCB1-AS1 FAM83E CGB8 CGB7 SYNGR4 ADM5 CCDC114 BCL2L12 SNAR-C2 PLA2G4C-AS1 FGF21 CPT1C CGB3 BCAT2 TMEM143 CARD8 FTL MIR5088 CYTH2 MIR4751 IL4I1 MIR6798 MIR4749 SLC17A7 LHB NOSIP NTN5 DKKL1 HRC RASIP1 LOC100287477 SNAR-A2 ATF5 SNAR-A9 LIN7B SNAR-C3 IRF3 FLT3LG BAX DHDH VRK3 SNORD35A SNAR-B2 TBC1D17 ELSPBP1 SPHK2 SNAR-C4 SNRNP70 CD37 |
nsv470150 | Chr.19:49618821 - 49856789 on Build GRCh38 | Gain | TSKS SCAF1 PRR12 PRMT1 ADM5 BCL2L12 PTOV1-AS1 CPT1C AP2A1 RRAS IRF3 MIR6799 MED25 PTOV1 MIR6800 MIR5088 PTOV1-AS2 MIR4749 FUZ |
dgv53n68 | Chr.19:49724393 - 49943224 on Build GRCh38 | Loss | TSKS AKT1S1 PNKP PTOV1-AS1 AP2A1 ATF5 NUP62 MIR6799 MED25 PTOV1 MIR6800 MIR4750 PTOV1-AS2 MIR4751 IL4I1 TBC1D17 MIR4749 FUZ |
nsv953600 | Chr.19:49555044 - 49938043 on Build GRCh38 | Deletion | TSKS ADM5 AKT1S1 BCL2L12 PNKP CPT1C AP2A1 PRRG2 MED25 MIR6800 MIR5088 MIR4750 PTOV1-AS2 MIR4751 IL4I1 MIR4749 SCAF1 NOSIP PRR12 PRMT1 PTOV1-AS1 ATF5 NUP62 RRAS IRF3 MIR6799 PTOV1 TBC1D17 FUZ |
nsv1160655 | Chr.19:49489110 - 49841830 on Build GRCh38 | Deletion | TSKS SNORD33 ADM5 BCL2L12 CPT1C RPS11 AP2A1 MIR150 PRRG2 SNORD32A MED25 MIR6800 MIR5088 RCN3 FCGRT SNORD34 SCAF1 RPL13A NOSIP PRR12 PRMT1 PTOV1-AS1 SNORD35B RRAS IRF3 MIR6799 SNORD35A FUZ |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |