Assay Details
Target Gene Details
Entrez Gene ID: | 5064 |
Gene Name: | paralemmin |
Gene Aliases: |
- |
Location: |
Chr.19:708953-748330 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PALM | NM_001040134.1 | NP_001035224.1 | ||
NM_002579.2 | NP_002570.2 | |||
XM_005259565.4 | XP_005259622.1 | |||
XM_005259566.4 | XP_005259623.1 | |||
XM_017026850.1 | XP_016882339.1 | |||
AK299809.1 | ||||
AK315168.1 | ||||
BC032449.1 | AAH32449.1 | |||
DA521203.1 | ||||
Y14770.1 | CAB37400.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv134n111 | Chr.19:607984 - 967584 on Build GRCh38 | Deletion | RNU6-9 CFD PALM ELANE MIR3187 FSTL3 MIR4745 MISP LOC101928450 HCN2 KISS1R RNF126 MED16 PRTN3 PRSS57 FGF22 POLRMT R3HDM4 PLPPR3 PTBP1 ARID3A AZU1 |
nsv577978 | Chr.19:294526 - 782854 on Build GRCh38 | Gain | SHC2 THEG PALM TPGS1 ODF3L2 BSG CDC34 FSTL3 MIER2 MISP LOC101928450 HCN2 LOC101929060 RNF126 GZMM C2CD4C PRSS57 FGF22 POLRMT MADCAM1 |
nsv953938 | Chr.19:355801 - 1104501 on Build GRCh38 | Deletion | SHC2 CFD GPX4 TPGS1 BSG ELANE FSTL3 LOC101928450 CNN2 PRTN3 FGF22 PLPPR3 ARHGAP45 PTBP1 RNU6-9 TMEM259 THEG PALM GRIN3B LOC105372235 ODF3L2 CDC34 WDR18 MIR3187 MIR4745 MISP HCN2 RNU6-2 KISS1R RNF126 POLR2E GZMM MED16 C2CD4C PRSS57 ABCA7 POLRMT R3HDM4 MADCAM1 ARID3A AZU1 |
nsv470101 | Chr.19:401714 - 1008505 on Build GRCh38 | Loss | SHC2 CFD TPGS1 BSG ELANE FSTL3 LOC101928450 PRTN3 FGF22 PLPPR3 PTBP1 RNU6-9 PALM GRIN3B LOC105372235 ODF3L2 CDC34 WDR18 MIR3187 MIR4745 MISP HCN2 KISS1R RNF126 GZMM MED16 C2CD4C PRSS57 POLRMT R3HDM4 MADCAM1 ARID3A AZU1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |