Assay Details
Target Gene Details
Entrez Gene ID: | 5064 |
Gene Name: | paralemmin |
Gene Aliases: |
- |
Location: |
Chr.19:708953-748330 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PALM | NM_001040134.1 | NP_001035224.1 | ||
NM_002579.2 | NP_002570.2 | |||
XM_005259565.4 | XP_005259622.1 | |||
XM_005259566.4 | XP_005259623.1 | |||
XM_017026850.1 | XP_016882339.1 | |||
AK299809.1 | ||||
AK315168.1 | ||||
BC032449.1 | AAH32449.1 | |||
DA521203.1 | ||||
Y14770.1 | CAB37400.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv134n111 | Chr.19:607984 - 967584 on Build GRCh38 | Deletion | FGF22 RNU6-9 HCN2 ARID3A PRSS57 MIR3187 MISP POLRMT PALM MED16 FSTL3 ELANE MIR4745 R3HDM4 PRTN3 CFD LOC101928450 AZU1 PLPPR3 KISS1R PTBP1 RNF126 |
nsv577978 | Chr.19:294526 - 782854 on Build GRCh38 | Gain | FGF22 TPGS1 HCN2 PRSS57 MISP POLRMT PALM FSTL3 ODF3L2 THEG LOC101928450 SHC2 BSG CDC34 LOC101929060 GZMM MADCAM1 C2CD4C MIER2 RNF126 |
nsv953938 | Chr.19:355801 - 1104501 on Build GRCh38 | Deletion | FGF22 RNU6-9 WDR18 ARID3A PRSS57 MIR3187 MISP POLRMT PALM ABCA7 ELANE MIR4745 GRIN3B PRTN3 CFD LOC105372235 THEG CDC34 PLPPR3 MADCAM1 POLR2E C2CD4C TPGS1 CNN2 HCN2 MED16 TMEM259 FSTL3 R3HDM4 RNU6-2 GPX4 ODF3L2 LOC101928450 SHC2 BSG AZU1 GZMM KISS1R PTBP1 ARHGAP45 RNF126 |
nsv470101 | Chr.19:401714 - 1008505 on Build GRCh38 | Loss | FGF22 RNU6-9 WDR18 ARID3A PRSS57 MIR3187 MISP POLRMT PALM ELANE MIR4745 GRIN3B PRTN3 CFD LOC105372235 CDC34 PLPPR3 MADCAM1 C2CD4C TPGS1 HCN2 MED16 FSTL3 R3HDM4 ODF3L2 LOC101928450 SHC2 BSG AZU1 GZMM KISS1R PTBP1 RNF126 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |