Assay Details
Target Gene Details
Entrez Gene ID: | 100128675 |
Gene Name: | HPN antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:35059059-35106304 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HPN-AS1 | NR_024562.1 | |||
AK295317.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)