Assay Details
Target Gene Details
Entrez Gene ID: | 23526 |
Gene Name: | Rho GTPase activating protein 45 |
Gene Aliases: |
HA-1, HLA-HA1, HMHA1 |
Location: |
Chr.19:1065923-1087831 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARHGAP45 | NM_001258328.2 | NP_001245257.1 | ||
NM_001321232.1 | NP_001308161.1 | |||
NM_012292.4 | NP_036424.2 | |||
XM_006722713.1 | XP_006722776.1 | |||
XM_011527858.1 | XP_011526160.1 | |||
AK300341.1 | ||||
AK304271.1 | ||||
AK304765.1 | ||||
BC035564.1 | AAH35564.1 | |||
BC065223.1 | AAH65223.1 | |||
D86976.1 | ||||
DB135747.1 | ||||
DB148385.1 | ||||
DB162560.1 | ||||
DC382305.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1058677 | Chr.19:1054061 - 1283189 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv136n111 | Chr.19:1038223 - 1329640 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv9662 | Chr.19:893394 - 1317740 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1160555 | Chr.19:887275 - 1346084 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3643383 | Chr.19:1012555 - 1069137 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv470102 | Chr.19:1046521 - 1152656 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv469845 | Chr.19:951640 - 1144486 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2758743 | Chr.19:951641 - 1544934 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv953938 | Chr.19:355801 - 1104501 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
dgv135n111 | Chr.19:1014378 - 1156398 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Additional Information:
For this assay, SNP(s) [rs113294002] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)