Assay Details
Target Gene Details
Entrez Gene ID: | 3991 |
Gene Name: | lipase E, hormone sensitive type |
Gene Aliases: |
AOMS4, FPLD6, HSL, LHS |
Location: |
Chr.19:42401507-42427426 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPE | NM_005357.3 | NP_005348.2 | ||
XM_005258937.3 | XP_005258994.1 | |||
XM_005258938.4 | XP_005258995.1 | |||
XM_005258939.3 | XP_005258996.2 | |||
XM_005258940.3 | XP_005258997.1 | |||
XM_006723218.2 | XP_006723281.1 | |||
XM_017026810.1 | XP_016882299.1 | |||
AK292482.1 | ||||
BC070041.1 | AAH70041.1 | |||
U40002.1 | AAC50666.1 |
Target Gene Details
Entrez Gene ID: | 100996307 |
Gene Name: | LIPE antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:42397148-42652355 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LIPE-AS1 | NR_073180.1 | |||
AK311181.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953584 | Chr.19:42387549 - 42412748 on Build GRCh38 | Deletion | LIPE-AS1 LIPE CNFN LOC101930071 |
nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss | ZNF574 TMEM145 LIPE GRIK5 LOC390937 POU2F2 LOC101930071 LOC100505585 CIC MIR8077 CD79A MIR6797 ERF ARHGEF1 LIPE-AS1 MEGF8 LOC100505622 GSK3A DEDD2 ZNF526 RABAC1 RPS19 CNFN PRR19 PAFAH1B3 MIR4323 ATP1A3 |
nsv526186 | Chr.19:42172329 - 42699355 on Build GRCh38 | Gain | LIPE-AS1 MEGF8 GSK3A TMEM145 LIPE DEDD2 CEACAM8 CEACAM1 POU2F2 LOC101930071 ZNF526 CIC MIR8077 CNFN PRR19 CXCL17 PAFAH1B3 ERF |
nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain | ZNF574 CEACAM7 TMEM145 LIPE GRIK5 LOC390937 POU2F2 LOC101930071 LOC100505585 CIC CEACAM6 MIR8077 DMRTC2 CD79A MIR6797 ERF CEACAM4 ARHGEF1 LIPE-AS1 MEGF8 LOC100505622 GSK3A DEDD2 ZNF526 RABAC1 LYPD4 RPS19 CNFN PRR19 PAFAH1B3 CEACAM3 MIR4323 CEACAM5 ATP1A3 |
esv2422471 | Chr.19:42228989 - 43165333 on Build GRCh38 | Deletion | LIPE-AS1 MEGF8 GSK3A TMEM145 LIPE LOC100289650 CEACAM8 PSG11 CEACAM1 PSG8 LOC101930071 PSG3 PSG2 PSG6 CIC MIR8077 PSG10P CNFN PSG7 PRR19 CXCL17 PAFAH1B3 ERF PSG1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |