Assay Details
Target Gene Details
Entrez Gene ID: | 10297 |
Gene Name: | APC2, WNT signaling pathway regulator |
Gene Aliases: |
APCL |
Location: |
Chr.19:1446268-1473244 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APC2 | NM_005883.2 | NP_005874.1 | ||
XM_005259475.2 | XP_005259532.1 | |||
XM_006722607.2 | XP_006722670.1 | |||
XM_006722608.3 | XP_006722671.2 | |||
XM_006722610.3 | XP_006722673.2 | |||
AF110334.1 | AAD28183.1 | |||
AL078616.1 | ||||
BC032573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv32942 | Chr.19:1320268 - 1999205 on Build GRCh38 | Gain+Loss | KLF16 CSNK1G2-AS1 GAMT MBD3 NDUFS7 DAZAP1 CSNK1G2 LOC100288123 C19orf25 MUM1 RPS15 SCAMP4 PCSK4 REXO1 BTBD2 ADAT3 MEX3D ATP8B3 REEP6 ABHD17A MIR1909 UQCR11 ADAMTSL5 APC2 ONECUT3 TCF3 PLK5 |
nsv578279 | Chr.19:1413575 - 1530061 on Build GRCh38 | Gain | RPS15 REEP6 PCSK4 ADAMTSL5 DAZAP1 C19orf25 PLK5 APC2 |
nsv833705 | Chr.19:1380125 - 1544934 on Build GRCh38 | Loss | RPS15 GAMT REEP6 PCSK4 ADAMTSL5 NDUFS7 DAZAP1 C19orf25 PLK5 APC2 |
esv2758743 | Chr.19:951641 - 1544934 on Build GRCh38 | Gain+Loss | CIRBP-AS1 C19orf24 GAMT POLR2E CIRBP LOC102725180 NDUFS7 DAZAP1 C19orf25 ATP5D MUM1 RPS15 TMEM259 ARHGAP45 ABCA7 PCSK4 CNN2 LOC105372235 REEP6 ADAMTSL5 CBARP APC2 GPX4 SBNO2 STK11 GRIN3B WDR18 ARID3A RNU6-2 EFNA2 MIDN PLK5 |
nsv953942 | Chr.19:1371602 - 2057401 on Build GRCh38 | Deletion | KLF16 CSNK1G2-AS1 GAMT MBD3 NDUFS7 DAZAP1 CSNK1G2 LOC100288123 C19orf25 MUM1 RPS15 SCAMP4 PCSK4 REXO1 BTBD2 ADAT3 MEX3D ATP8B3 REEP6 ABHD17A MIR1909 UQCR11 ADAMTSL5 APC2 ONECUT3 MKNK2 TCF3 PLK5 |
nsv470105 | Chr.19:1444420 - 1526614 on Build GRCh38 | Gain | REEP6 PCSK4 ADAMTSL5 C19orf25 PLK5 APC2 |
More Information
Additional Information:
For this assay, SNP(s) [rs80271587] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |