Assay Details
Target Gene Details
Entrez Gene ID: | 25759 |
Gene Name: | SHC adaptor protein 2 |
Gene Aliases: |
SCK, SHCB, SLI |
Location: |
Chr.19:416583-461011 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SHC2 | NM_012435.2 | NP_036567.2 | ||
XM_011527893.2 | XP_011526195.1 | |||
XM_011527894.2 | XP_011526196.1 | |||
XM_011527895.2 | XP_011526197.1 | |||
XM_011527896.2 | XP_011526198.1 | |||
XM_017026567.1 | XP_016882056.1 | |||
AB001451.1 | BAA25798.1 | |||
AL360254.1 | CAB96175.1 | |||
BY796046.2 | ||||
CF457169.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509712 | Chr.19:391096 - 558643 on Build GRCh38 | Insertion | GZMM C2CD4C CDC34 ODF3L2 TPGS1 MADCAM1 SHC2 |
dgv6155n54 | Chr.19:404077 - 519032 on Build GRCh38 | Loss | C2CD4C ODF3L2 TPGS1 MADCAM1 SHC2 |
nsv428360 | Chr.19:232069 - 702131 on Build GRCh38 | Gain | GZMM CDC34 RNF126 ODF3L2 THEG FSTL3 TPGS1 MIER2 LOC101929060 MADCAM1 SHC2 BSG C2CD4C POLRMT FGF22 HCN2 PRSS57 PLPP2 |
dgv1013e212 | Chr.19:416231 - 521269 on Build GRCh38 | Gain | ODF3L2 TPGS1 MADCAM1 SHC2 |
esv2671474 | Chr.19:393636 - 481273 on Build GRCh38 | Deletion | C2CD4C ODF3L2 SHC2 |
nsv1065558 | Chr.19:305266 - 441626 on Build GRCh38 | Gain | C2CD4C THEG MIER2 SHC2 |
nsv578091 | Chr.19:411849 - 431187 on Build GRCh38 | Gain | SHC2 |
nsv577978 | Chr.19:294526 - 782854 on Build GRCh38 | Gain | GZMM CDC34 RNF126 ODF3L2 THEG FSTL3 LOC101928450 TPGS1 MIER2 LOC101929060 MADCAM1 SHC2 BSG C2CD4C POLRMT FGF22 PALM MISP HCN2 PRSS57 |
nsv953938 | Chr.19:355801 - 1104501 on Build GRCh38 | Deletion | GZMM CDC34 ODF3L2 THEG FSTL3 LOC101928450 TPGS1 MIR3187 MED16 MIR4745 R3HDM4 MADCAM1 SHC2 ABCA7 C2CD4C PLPPR3 POLRMT RNU6-9 AZU1 FGF22 CFD HCN2 RNU6-2 TMEM259 RNF126 POLR2E CNN2 LOC105372235 WDR18 BSG ELANE ARID3A PTBP1 PRTN3 ARHGAP45 PALM GPX4 MISP PRSS57 GRIN3B KISS1R |
nsv470101 | Chr.19:401714 - 1008505 on Build GRCh38 | Loss | GZMM CDC34 ODF3L2 FSTL3 LOC101928450 TPGS1 MIR3187 MED16 MIR4745 R3HDM4 MADCAM1 SHC2 C2CD4C PLPPR3 POLRMT RNU6-9 AZU1 FGF22 CFD HCN2 RNF126 LOC105372235 WDR18 BSG ELANE ARID3A PTBP1 PRTN3 PALM MISP PRSS57 GRIN3B KISS1R |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |