Assay Details
Target Gene Details
Entrez Gene ID: | 79948 |
Gene Name: | phospholipid phosphatase related 3 |
Gene Aliases: |
LPPR3, LPR3, PRG-2, PRG2 |
Location: |
Chr.19:812488-824244 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLPPR3 | NM_001270366.1 | NP_001257295.1 | ||
NM_024888.2 | NP_079164.1 | |||
XM_011528317.2 | XP_011526619.1 | |||
AF357888.1 | AAO85401.1 | |||
AF541282.1 | AAP57771.1 | |||
AL136596.1 | CAB66531.1 | |||
AY304517.1 | AAP72154.1 | |||
AY436785.1 | AAR10818.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv828357 | Chr.19:817381 - 823632 on Build GRCh38 | Loss | PLPPR3 |
dgv134n111 | Chr.19:607984 - 967584 on Build GRCh38 | Deletion | HCN2 FGF22 R3HDM4 PTBP1 PRTN3 PRSS57 POLRMT CFD LOC101928450 PLPPR3 MISP RNU6-9 MIR4745 MIR3187 MED16 ARID3A AZU1 RNF126 FSTL3 ELANE PALM KISS1R |
nsv578172 | Chr.19:811414 - 841398 on Build GRCh38 | Loss | MIR3187 PTBP1 PRTN3 AZU1 PLPPR3 |
nsv578167 | Chr.19:797079 - 850574 on Build GRCh38 | Loss | MIR4745 MIR3187 PTBP1 PRTN3 AZU1 PLPPR3 |
nsv953938 | Chr.19:355801 - 1104501 on Build GRCh38 | Deletion | HCN2 FGF22 PTBP1 PRTN3 ABCA7 THEG SHC2 POLRMT GRIN3B PLPPR3 RNU6-9 MIR4745 POLR2E MIR3187 ARHGAP45 LOC105372235 RNF126 ODF3L2 PALM C2CD4C CDC34 R3HDM4 PRSS57 MADCAM1 BSG CFD LOC101928450 MISP MED16 ARID3A WDR18 CNN2 AZU1 GPX4 FSTL3 GZMM ELANE TMEM259 RNU6-2 TPGS1 KISS1R |
nsv470101 | Chr.19:401714 - 1008505 on Build GRCh38 | Loss | HCN2 FGF22 PTBP1 PRTN3 SHC2 POLRMT GRIN3B PLPPR3 RNU6-9 MIR4745 MIR3187 LOC105372235 RNF126 ODF3L2 PALM C2CD4C CDC34 R3HDM4 PRSS57 MADCAM1 BSG CFD LOC101928450 MISP MED16 ARID3A WDR18 AZU1 FSTL3 GZMM ELANE TPGS1 KISS1R |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |